Tlačiť

Anti-MYC Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB10424-100 ORB10424-200
BIRBORB10424-100EA 0 EUR
BIRBORB10424-100 BIRBORB10424-200
Anti-MYC Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR