Tlačiť

Anti-mu Opioid receptor Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB6275-200 ORB6275-100
BIRBORB6275-200EA 0 EUR
BIRBORB6275-200 BIRBORB6275-100
Anti-mu Opioid receptor Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: mu Opioid receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR