Tlačiť

Anti-PROK1 Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB5612-100 ORB5612-200
BIRBORB5612-100EA 0 EUR
BIRBORB5612-100 BIRBORB5612-200
Anti-PROK1 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: PROK1 (prokineticin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR