Tlačiť

Anti-TAAL6 Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB2084-100 ORB2084-200
BIRBORB2084-100EA 0 EUR
BIRBORB2084-100 BIRBORB2084-200
Anti-TAAL6 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TAAL6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR