Tlačiť

Anti-PAP alpha Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB1118-100 ORB1118-200
BIRBORB1118-100EA 0 EUR
BIRBORB1118-100 BIRBORB1118-200
Anti-PAP alpha Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: PAP alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR