Tlačiť
Peqlab, PCR Tubes
Dodávateľ: VWR Peqlab
732-3206EA 87.44 EUR
732-3206 732-3207 732-3215 732-3216
Peqlab, PCR Tubes
Zkumavky Zkumavky pro PCR
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Ultra-thin walls for rapid heat transfer
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.