Tlačiť
Capillary columns, Ultra 2
Dodávateľ: AGILENT HEWLETT PACKARD
19091B-101LTM 19091B-102LTM 19091B-112LTM
HEWL19091B-101LTMEA 0 EUR
HEWL19091B-101LTM HEWL19091B-102LTM HEWL19091B-112LTM
Capillary columns, Ultra 2
Kolony chromatografické
Columns equivalent to HP-5 with tighter specifications for retention index and capacity factors.
  • Non polar
  • Bonded and cross-linked
  • Solvent rinsable
(5%-Phenyl)-methylpolysiloxane