Tlačiť
Capillary columns, DB-1
Dodávateľ: AGILENT HEWLETT PACKARD
123-106B 12A-1015 121-1012LTM 121-1013LTM 121-101ALTM 121-1022LTM 121-1023LTM 122-1012LTM 122-1022LTM 122-1032LTM 122-1033LTM 122-103ELTM 123-1011LTM 123-1012LTM 123-1015LTM 123-1033LTM 123-1035LTM 123-103BLTM 123-103ELTM 127-100ALTM 127-1013LTM 127-1023LTM 127-1046E 128-1012LTM 128-1022LTM 12A-1015LTM
HEWL123-106BEA 1902.93 EUR
HEWL123-106B HEWL12A-1015 HEWL121-1012LTM HEWL121-1013LTM HEWL121-101ALTM HEWL121-1022LTM HEWL121-1023LTM HEWL122-1012LTM HEWL122-1022LTM HEWL122-1032LTM HEWL122-1033LTM HEWL122-103ELTM HEWL123-1011LTM HEWL123-1012LTM HEWL123-1015LTM HEWL123-1033LTM HEWL123-1035LTM HEWL123-103BLTM HEWL123-103ELTM HEWL127-100ALTM HEWL127-1013LTM HEWL127-1023LTM HEWL127-1046E HEWL128-1012LTM HEWL128-1022LTM HEWL12A-1015LTM
Capillary columns, DB-1
Kolony chromatografické
These columns contain 100% dimethylpolysiloxane.
  • Non-polar
  • Very low bleed characteristics, ideal for GC/MS
Max. temperature: -60 to 325/350 °C