Tlačiť
Capillary columns, DB-5
Dodávateľ: Agilent
J&W
HEWL123-500ELTMEA 1995.23 EUR
HEWL123-5033 HEWL123-500ELTM HEWL121-5022 HEWL121-5023 HEWL123-5031 HEWL123-5032 HEWL125-5065E HEWL123-5011LTM HEWL123-503E HEWL123-503B HEWL122-5022 HEWL125-503D HEWL125-503B HEWL128-50H7 HEWL125-503K HEWL121-5012 HEWL125-503J HEWL122-503E HEWL121-5013 HEWL122-5031 HEWL125-5034 HEWL121-5012LTM HEWL125-5032 HEWL12A-5015LTM HEWL125-5037 HEWL122-5033 HEWL125-5035 HEWL122-5032 HEWL123-5033E HEWL121-5023LTM HEWL123-5056 HEWL122-5032E HEWL123-5052 HEWL123-5053 HEWL121-5042 HEWL123-5032LTM HEWL123-5013LTM HEWL125-5012E HEWL125-501K HEWL123-503BLTM HEWL125-5025 HEWL123-5032E HEWL127-501ELTM HEWL122-5033E HEWL127-5012LTM HEWL124-5032 HEWL123-5062 HEWL123-5063 HEWL125-501J HEWL121-5022E HEWL124-5037 HEWL122-501E HEWL122-5033LTM HEWL128-5012 HEWL125-5012 HEWL123-503ELTM HEWL127-5022E HEWL125-5017 HEWL122-5002LTM HEWL122-5013 HEWL122-5012 HEWL122-5011 HEWL127-501E HEWL12A-5015 HEWL122-506E HEWL122-5063 HEWL122-5062 HEWL122-5061 HEWL122-5032LTM HEWL125-50HB HEWL128-5022 HEWL127-5013 HEWL127-5012 HEWL121-5022LTM HEWL122-5063E HEWL123-5012LTM HEWL121-5012E HEWL123-500E HEWL125-5035E HEWL127-5012E HEWL122-5062E HEWL127-5013LTM HEWL127-5023 HEWL127-5022 HEWL127-501N HEWL122-503ELTM HEWL122-5012E HEWL123-5011 HEWL123-5013E HEWL123-5012 HEWL123-5013 HEWL121-5013LTM HEWL125-5065 HEWL125-5062 HEWL123-5003LTM HEWL123-5026 HEWL123-5012E HEWL123-5022 HEWL125-5035LTM HEWL125-5032E HEWL123-502F HEWL122-5052 HEWL128-5052 HEWL122-5012LTM HEWL123-502D HEWL125-5032LTM
Capillary columns, DB-5
Kolony chromatografické
These non-polar columns are used in wide range of applications.
  • Low bleed, bonded and cross-linked
  • Solvent rinsable
  • High temperature limit
(5%-Phenyl)-methylpolysiloxane.