Tlačiť
Cleanroom finger cots, nitrile
Dodávateľ: Basan
380041 380042
BSNC380041EA 0 EUR
BSNC380041 BSNC380042
Cleanroom finger cots, nitrile
Návleky na prsty Návleky na prsty nitrilové
These disposable, rolled finger cots are made from 100% nitrile. They are recommended for use when handling electrostatic sensitive devices or ceramic components in areas such as ESD, aviation, marine, electronics, jewelry industry, repair stations and assembly areas.
  • Anti-static, powder-free
  • Completely free of latex and latex proteins
  • Surface resistivity: ≤10¹⁰ ohms/sq
Suitable for use in Class ISO 5, 100 or M3.5 cleanrooms.
Thickness: 0,09 mm
Colour: White
Balenie: Washed and packed in cleanroom environment. Double packed in bag.