Tlačiť
InGel™ Blue for protein digestion
Dodávateľ: G-Biosciences
786-681EA 550.04 EUR
786-681
InGel™ Blue for protein digestion
Systémy pro extrakci proteinů Činidla pro extrakci bílkovin
InGel™ Blue provides a complete set of reagents for the in-gel tryptic digestion and extraction of peptides for mass spectrometry (MALDI and LC MS/MS). The kit is specifically designed for use with Coomassie® or fluorescent stained protein spots/bands.
  • For the in-gel tryptic digestion of proteins
  • Compatible with Coomassie® and fluorescent stained proteins
  • Supplied with mass spectrometry grade trypsin
  • Supplied with destaining, reducing, alklating and peptide extraction reagents