Tlačiť
Detergent removal system, DetergentOUT™ Tween®
Dodávateľ: G Bioscienes
786-214 786-215
786-214EA 534.7 EUR
786-214 786-215
Detergent removal system, DetergentOUT™ Tween®
Systémy pro extrakci proteinů Činidla pro extrakci bílkovin
DetergentOUT™ Tween® is a spin column format detergent removal resin for polysorbate or Tween® detergents or surfactants.
DetergentOUT™ Tween® specifically removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d52c1043214149edc1c697a3ffffe58f6c025f&vid=0633c24839d746937941c948790c95906898733e&prid=web&clid=SAPDAM