Tlačiť
ESD safety shoes, lace-up, uvex 1 ladies, 8561
Dodávateľ: UVEX
8561.8/36 8561.8/43 8561.8/42 8561.8/41 8561.8/40 8561.8/39 8561.8/37 8561.8/35 8561.8/38
UVEX8561.8/36EA 156.44 EUR
UVEX8561.8/36 UVEX8561.8/43 UVEX8561.8/42 UVEX8561.8/41 UVEX8561.8/40 UVEX8561.8/39 UVEX8561.8/37 UVEX8561.8/35 UVEX8561.8/38
ESD safety shoes, lace-up, uvex 1 ladies, 8561
Obuv
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Grey/pink
Width: 11
Certifikácie: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC