Tlačiť
Western blots, pre-made, human single tissue single species
Dodávateľ: G Bioscienes
TB01 TB04 TB07 TB10 TB13 TB16 TB19 TB22 TB25 TB54 TB88 TB74 TB75 TB76 TB77 TB78 TB80 TB81 TB82 TB83 TB84 TB85 TB87 TB71 TB89 TB90 TB91 TB92 TB93 TB94 TB95 TB96 TB97 TB98 TB86 TB100 TB73 TB72 TB60 TB61 TB62 TB63 TB64 TB65 TB66 TB67 TB68 TB69 TB70
TB01EA 806.26 EUR
TB01 TB04 TB07 TB10 TB13 TB16 TB19 TB22 TB25 TB54 TB88 TB74 TB75 TB76 TB77 TB78 TB80 TB81 TB82 TB83 TB84 TB85 TB87 TB71 TB89 TB90 TB91 TB92 TB93 TB94 TB95 TB96 TB97 TB98 TB86 TB100 TB73 TB72 TB60 TB61 TB62 TB63 TB64 TB65 TB66 TB67 TB68 TB69 TB70
Western blots, pre-made, human single tissue single species
Činidla pro elektroforézu Western Blotting Reagents
Ready-to-screen, human, single tissue blots, from both normal and tumour tissues. G-Biosciences pre-made blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Western transfer buffer.
G-Biosciences pre-made blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Objednávacie informácie: Accessories available.