Tlačiť
Sample containers, with screw cap, non-pyrogenic, Sterilin®
Dodávateľ: THERMO STERILIN
128PYR 125PYR 129PYR 190PYR 165PYR 185PYR
215-3684EA 784.6 EUR
215-3684 215-3683 216-1558 215-3687 215-3685 215-3686
Sample containers, with screw cap, non-pyrogenic, Sterilin®
Nádobky na vzorky Nádoby pro obecné použití
PS, with PE, PP oder metal screw cap.
  • In vitro use only
  • Gamma irradiated
  • Leak tested in accordance with EN14254 Annex D and BS5213
  • Lot number on each container label and test certificate supplied for each lot
Suitable for the storage of samples/formulation work/ sterility testing and endotoxin testing. Certified endotoxin free to levels below 0,01 EU/ml (Limulus Amoebocyte Lysate (LAL) test).
Perform sterility and endotoxin tests with Thermo Scientific™ Sterilin™ non-pyrogenic polystyrene containers. Not available in America.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904