Tlačiť
VWR®, Test Sieves, 315×75 mm, with Square Holes
Dodávateľ: VWR Collection
510-1187EA 346.08 EUR
510-1187 510-1188 510-1189 510-1190 510-1191 510-1192 510-1193 510-1194 510-1195 510-1196 510-1197 510-1198 510-1199 510-1200 510-1202 510-1203 510-1204 510-1205 510-1206 510-1207 510-1208 510-1209 510-1210 510-1211 510-1212 510-1213 510-1214 510-1215 510-1216 510-1217 510-1218 510-1219 510-1220 510-1221 510-1222 510-1223 510-1224 510-1225 510-1226 510-1227 510-1228
VWR®, Test Sieves, 315×75 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.