Tlačiť
Balance boats, disposable
Dodávateľ: KARTELL
11103 11104 11105 1110003 1110103 1110109 1110203 1110209
KART11103EA 215.4 EUR
KART11103 KART11104 KART11105 KART1110003 KART1110103 KART1110109 KART1110203 KART1110209
Balance boats, disposable
Lodičky Lodičky navažovací
Balance boats, high impact PS.
  • Easily fashioned to make a funnel shape
  • The DS model is available in either white or black so the operator can easily see that no trace of sample remains adhering to the boat
  • Reproducible tare weight: batch to batch variation is normally less than 5%
  • Antistatic
Diamond (DS) and square shaped (SQ) lightweight weighing boats provide an easy and inexpensive system for weighing any medium with non contaminating properties (made of HIPS with hydrophobic surface).