Tlačiť
PCR plates, 384-well, opaque, skirted, Amplate™
Dodávateľ: SIMPORT SCIENTIFIC
T324-384SKK T324-384SKW
SIMPT324-384SKKEA 527.08 EUR
SIMPT324-384SKK SIMPT324-384SKW
PCR plates, 384-well, opaque, skirted, Amplate™
Mikrodestičky Destičky pro PCR
PP. These opaque 384-well PCR plates are for chemiluminescent and fluorescent procedures. Thin-walled and designed for rapid thermal transfer, these plates are precision-moulded to ensure well-to-well and plate-to-plate uniformity.
  • Opaque to ensure low level of background fluorescence
  • Can be filled using automated fluid handling systems
  • Two corners are cut away to facilitate orientation
  • Alphanumeric grid for better identification
  • Can be handled by robotic handling equipment
  • Opaque to ensure low level of background fluorescence
  • Area for bar coding, labelling or writing
Informace o dodávce: Packed in tamperproof resealable bags of 10 plates.