Tlačiť
Surface abrasives, Polish-fix
Dodávateľ: BOCHEM
8901 8902 8903
149-1911EA 20.42 EUR
149-1911 149-1912 149-1913
Surface abrasives, Polish-fix
Prostředky čistící Detergenty a povrchové čističe
Abrasive cleaners, cleans, polishes and matt-finishes.
  • Type 1: grain size 60, rough
  • Type 2: grain size 120, medium
  • Type 3: grain size 240, fine
For dry and wet application with water, solvents or detergents. Suitable for jobs around the house, workshop and leisure, for working metal, paints, wood, leather and rubber.