Tlačiť
Labelling tapes, adhesive, NeoTape®
Dodávateľ: NEOLAB
2-6108 2-6109 2-6110 2-6114
NEOL2-6108EA 14.38 EUR
NEOL2-6108 NEOL2-6109 NEOL2-6110 NEOL2-6114
Labelling tapes, adhesive, NeoTape®
Pásky
Universal coloured self-adhesive label tapes are suitable for innumerable applications in laboratories, workshops, hospital rooms, doctor's practices, and more.
  • Can be written on with pencil, pen, felt-pen, ink, typewriter etc
  • Removes without residue and may be used for second time
  • Oil, water and acid resistant
  • Withstands temperatures from −45 to +121 °C