Tlačiť
Blood collection tubes, with ACD, VACUETTE®
Dodávateľ: Greiner Bio-One
455094_1200 456094_1200 455055_1200
GREI455094_1200EA 296.83 EUR
GREI455094_1200 GREI456094_1200 GREI455055_1200
Blood collection tubes, with ACD, VACUETTE®
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These tubes are available with ACD solutions and are used for blood grouping determinations and also for cell preservation.