Tlačiť
Peha-Soft® Nitrile Nitrile Gloves, Sterile
Dodávateľ: HARTMANN PAUL
9422111 9422121 9422131
HART9422111EA 142.8 EUR
HART9422111 HART9422121 HART9422131
Peha-Soft® Nitrile Nitrile Gloves, Sterile
Rukavice
These latex-free disposable examination and protective gloves are made of soft, blue nitrile rubber.
  • Sterile, powder-free
  • The chlorinated inner lining makes the gloves easy to put on
  • Textured fingers for optimum grip
  • Highly elasticated, very tear-resistant and secure
  • Excellent fit and outstanding tactile sensitivity
  • Both left and right-handed
Ideal for situations in which staff are at risk of contamination, e.g. when changing dressings, catheterising or intubating. Particularly suited to people who are sensitive to glove powder or allergic to latex proteins (Type 1 allergy). Also suitable for laboratory work and cleaning with chemicals or disinfectants.
Colour: blue
EN 374, EN 455, Cat. III
Certifikácie: EN 374, EN 455, Cat. III