Tlačiť
Microbalances, BM series
Dodávateľ: AnD Instruments Ltd (A&D)
BM-20 BM-22
611-2857EA 0 EUR
611-2857 611-2858
Microbalances, BM series
Váhy
A highly specified range of microbalances from A&D with an upper neutralising chamber that houses a built in ioniser to eliminate static charge on the sample or sample container. The balance is supplied with a data logger that records up to 5000 results with time and date providing data not only about the weighing operation but also the environmental readings from built-in sensors.
  • Easy, fanless static elimination using a built-in ioniser
  • Double-ring structure around the weighing pan reduces effects of any localised draughts
  • Interlockable sliding doors allowing efficient operation with both hands
  • Data memory function for 100 weighing results with time and date
  • Built-in temperature, humidity, and atmospheric pressure sensors
  • Automatic calibration with an internal mass and automatic response adjustment
GLP/GMP/GCP/ISO compliant output
Certifikácie: GLP/GMP/GCP/ISO compliant output
Informace o dodávce: Balance supplied with WinCT (CD-ROM), weighing data logger, weighing pans and supports, tweezers and two larger (50 and 95 mm) weighing support pans for filter weighing, microtube holder and three sizes (8, 12 and 15 mm) of aluminium analytical pans (10 pcs each) AC adapter, USB cable, display cover and instruction manual.
Pozor: Please note these products are not available in Republic of Ireland or Northern Ireland.