Tlačiť
Glove liners, Puri Top Fit
Dodávateľ: PURITECH
TFG600-S TFG600-M TFG600-L TFG600-XL TFG600-XXL TFG600-XS TFG600XSINDPAC TFG600SINDPACK TFG600MINDPACK TFG600LINDPACK TFG600XLINDPAC TFG6002XLINDPC
112-3427EA 47.33 EUR
112-3427 112-3426 112-3425 112-3428 112-3430 112-3429 112-4268 112-4190 112-4189 112-4188 112-4191 112-4267
Glove liners, Puri Top Fit
Rukavice
These knitted nylon gloves feature a thin PU coating on the fingertips.
  • Breathable fabric
  • Optimum comfort and dexterity
  • Hand-specific for good fit
  • Washable at 40 °C
The gloves are ideal for handling small components, dry assembly applications, maintenance, pharmaceutical processing and other light duty applications. Suitable for use in ISO Class 7 and 8 cleanrooms.
Colour: White