Tlačiť
Biocide, Klercide™ low residue quat
Dodávateľ: ECOLAB CONTAMINATION CONTROL
3078730 3078740
141-1084EA 1904.93 EUR
141-1084 141-1083
Biocide, Klercide™ low residue quat
Prostředky čistící Dezinfekční prostředek
Klercide Low Residue Quat Blended with WFI version is a solution with water for injection (WFI) and the pack is triple bagged. Low endotoxin levels are guaranteed.
  • Sterile cleanroom biocide for all types of hard surfaces
  • Effective at low concentrations and non corrosive
  • Fully compatible and ideal for a rotational regime
  • SteriShield Delivery System™ ensures 3 months in-use shelf life
Suitable for ISO Class 5; Grade A and B (microbial count) areas: cleanroom processed, sterile filled and double bagged. Solution of quaternary ammonium compound and water, with broad spectrum activity. Filtered to 0,2 µm.
Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697.
Certifikácie: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697.
Pozor: Use biocides safely. Always read the label and product information before use