Tlačiť
High capacity balances, GF-K and GX-K series
Dodávateľ: AnD Instruments Ltd (A&D)
GF-10K GF-30K GX-32K GX-8K2
611-3075EA 0 EUR
611-3075 611-3078 611-3106 611-3108
High capacity balances, GF-K and GX-K series
Váhy
High capacity top loading balances from A&D featuring high speed weighing and settling times. Applications include counting, percentage weighing, animal weighing, below balance weighing and density determination. All models feature display auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement (ACAI). RS232C interface for printer or computer for GLP data output format to suit quality systems.
  • Rugged aluminium alloy enclosure and stainless steel pan with IP 65 waterproof and dust protection
  • Weighing mechanism Super Hybrid Sensor (SHS) gives fast response speed - typically just 1,5 seconds
  • Multiple weighing units plus counting, percentage and user programmable functionality
  • Automatically adjusts the environmental settings for GX models
  • Built-in under hook capability and animal weighing software
The GX-K balances store 200 sets of weighing data (or 100 sets of weighing data with time and date, or 50 sets of GLP data, or 20 sets of unit weight for counting mode, or upper and lower limit for comparator mode, or tare value).
GF-K models can store 40 sets of weighing data or 50 sets of unit weight for counting mode, or 20 sets of upper and lower limit values for comparator mode, or 20 sets of tare value.
Objednávacie informácie: Please contact your local Avantor sales office for details of accessories including animal weighing pan, draught shields, density determination kits and calibration weights and waterproof RS232 cable.
Informace o dodávce: Supplied with AC adapter and Windows Communication Tools (WinCT) data collection software.
Pozor: Please note these products are not available in Republic of Ireland or Northern Ireland.