Tlačiť
Precision balances, EK-i series
Dodávateľ: AnD Instruments Ltd (A&D)
EK-1200I EK-12KI EK-2000I EK-200I EK-3000I EK-300I EK-6100I-EC EK-610I-EC EK-6000I
611-2976EA 0 EUR
611-2976 611-2977 611-2978 611-2979 611-3016 611-3017 611-3022 611-3023 611-3020
Precision balances, EK-i series
Váhy
Compact, lightweight, low profile balances from A&D with a rugged housing and stainless steel pan for reliable, accurate weighing. External calibration with full digital calibration and user definable mass. The sealed key panel protects against dust and spills. All models feature auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement (ACAI). GLP conformity with ID and serial number outputs.
  • Nine built-in weighing units plus counting and percentage software
  • Large backlit LCD display with wide viewing angle
  • RS232C bi-directional interface
  • Optional rechargeable battery pack for portable operation
  • Levelling feet and level bubble for accurate weighing
Please note these products are not available in Republic of Ireland or Northern Ireland.
Objednávacie informácie: Please contact your local Avantor sales office for details on other accessories, including rechargeable Ni-MH battery, calibration weights and under balance weighing hook (for models EK-4100i /6100i /6000i /12Ki only).
Informace o dodávce: Supplied with AC adapter.