Tlačiť
High capacity balances, GP series
Dodávateľ: AnD Instruments Ltd (A&D)
GP-12K-EC GX22001L GP-32K-EC GP-60K-EC GP-100K-EC GP-102K-EC GP-100KS GP-30KS GP-32KS GP-60KS GP-61K GP-61KS
611-3086EA 0 EUR
611-3086 611-3087 611-3091 611-3094 611-3083 611-3085 611-3084 611-3089 611-3092 611-3095 611-3096 611-3097
High capacity balances, GP series
Váhy
The GP series of high capacity, high resolution balances from A&D are ideal for industrial applications. Units have a large stainless steel weighing pan and the display is mounted on a swing arm ('K' models) or detached ('KS' models). Applications include counting, density determination, percentage and animal weighing.
  • Multiple weighing units (including one custom unit)
  • Internal calibration using motor driven weights
  • GLP compliance with standard time and date function together with balance ID
  • Clear and large vacuum fluorescent display elements
  • RS232C bi-directional interface
All models feature display auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement (ACAI). The automatic self calibration feature responds to changes in ambient temperature.
Models denoted '-EC' are OIML EC approved, please contact your local Avantor sales office for full details.
Models denoted '-EC' are OIML EC approved, please contact VWR for full details.
Certifikácie: GP series balances are IP 65 compliant: Water- and dust-proof.
Objednávacie informácie: Please contact your local Avantor sales office for details on other accessories including animal weighing pan, under balance weighing hook, footswitch and calibration weights.
Informace o dodávce: Supplied with AC adapter and WinCT communication software.
Pozor: Please note these products are not available in Republic of Ireland or Northern Ireland.