Tlačiť
ESD safety shoes, sandals, Velcro® fastening, X-Light 7131031/7131036
Dodávateľ: Abeba
7131031/35 7131031/36 7131031/37 7131031/38 7131031/39 7131031/40 7131031/41 7131031/42 7131031/43 7131031/44 7131031/45 7131031/46 7131031/47 7131031/48 7131036/35 7131036/36 7131036/37 7131036/38 7131036/39 7131036/40 7131036/41 7131036/42 7131036/43 7131036/44 7131036/45 7131036/46 7131036/47 7131036/48
ABEB7131031/35EA 119.94 EUR
ABEB7131031/35 ABEB7131031/36 ABEB7131031/37 ABEB7131031/38 ABEB7131031/39 ABEB7131031/40 ABEB7131031/41 ABEB7131031/42 ABEB7131031/43 ABEB7131031/44 ABEB7131031/45 ABEB7131031/46 ABEB7131031/47 ABEB7131031/48 ABEB7131036/35 ABEB7131036/36 ABEB7131036/37 ABEB7131036/38 ABEB7131036/39 ABEB7131036/40 ABEB7131036/41 ABEB7131036/42 ABEB7131036/43 ABEB7131036/44 ABEB7131036/45 ABEB7131036/46 ABEB7131036/47 ABEB7131036/48
ESD safety shoes, sandals, Velcro® fastening, X-Light 7131031/7131036
Obuv
ESD safety shoe made from smooth leather, Air Mesh textile inlays, reflective material and a fuel resistant PU sole.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable active comfort insole provides optimised comfort; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Double Velcro® fastening
Colour: White or black
Colour: White/black
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S1, SRC.