Tlačiť
VWR® Q Path® MicroStar II, Cassettes, in Dispenser
Dodávateľ: VWR Collection
720-2382EA 274.38 EUR
720-2382 720-2383 720-2384 720-2385 720-2386 720-2387 720-2388 720-2389 720-2390
VWR® Q Path® MicroStar II, Cassettes, in Dispenser
Kazety pro zalévání vzorků Kazety bioptické
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with tape (= rack) for Leica and Sakura cassette printers and separate disposable lid, supplied in racks of 40 cassettes.
  • Small opening for the filtration of fluids
  • Hinge and separate lid
  • 45° writing angle