Tlačiť
IP ActivFlo Biopsy I cassettes, taped
Dodávateľ: LEICA HISTOLOGY
39LC-605-1-L 39LC-605-9-L 39LC-605-7-L 39LC-605-10-L 39LC-605-8-L 39LC-605-11-L 39LC-605-2-L 39LC-605-3-L 39LC-605-4-L 39LC-605-5-L 39LC-605-6-L
720-2362EA 202.97 EUR
720-2362 720-2650 720-2646 720-2492 720-2648 720-2494 720-2496 720-2498 720-2640 720-2642 720-2644
IP ActivFlo Biopsy I cassettes, taped
Kazety pro zalévání vzorků Kazety bioptické
IP ActivFlo Biopsy I cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes feature sloped walls, creating a basket for small tissue management. These one piece cassettes with ‘frangible lids’ have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Available for easy printer loading
  • Eleven colours in quantities of 1000 taped