Tlačiť
Electrical resistant glove liners, A0150 / A0160 / A0170
Dodávateľ: SHOWA BEST GLOVE
AO170-L AO160-M AO160-L AO160-XL AO160-S AO170-M AO170-XL AO170-S
112-4215EA 108.7 EUR
112-4215 112-4212 112-4211 112-4214 112-4213 112-4216 112-4218 112-4217
Electrical resistant glove liners, A0150 / A0160 / A0170
Rukavice
These low linting nylon/acrylic knitted gloves have anti-static properties. The conductive Thunderon® fibre covers the important area of the fingers, providing static discharge by corona effect. The gloves are available without coating, with PU coating on fingertips only, and with coating on fingertips and palm.
  • Surface resistivity of 10⁶ to 10⁷ Ω
  • Optimal dexterity and tactile feel
  • Seamless knit: No skin irritation
  • Elasticated cuffs: Great freedom of movement for the wrist
  • PU coating protects extremities from oils and micro cuts and prevents fingerprints on handled objects
Ideal for use when working with electronic products, in manufacturing industries and in automotive-related industries.
Colour: Light grey/white
A0150/A0160: EN 388 (0130); Cat. II A0170: EN 388 (3121); Cat. II
Certifikácie: A0150: EN 388:2016 (0130X); Cat. II
A0160: EN 388:2016 (0130X), EN 16350:2014; Cat. II
A0170: EN 388:2016 (3121X), EN 16350:2014; Cat. II