Tlačiť
Valve supplies for HPLC system
Dodávateľ: AGILENT HEWLETT PACKARD
0100-1853 5067-4117 5068-0006
HEWL0100-1853EA 242.28 EUR
HEWL0100-1853 HEWL5067-4117 HEWL5068-0006
Valve supplies for HPLC system
Chromatografické potřeby Chromatography Spare Parts
Vespel is a polyimide with low wear and high chemical resistance. Vespel tolerates a pH range of 0 to 10.
PEEK offers a high chemical resistance and flexibility and will tolerate the entire pH range from 0 to 14.
Tefzel is ideal for use in applications where PEEK cannot be used, such as methylene chloride or DMSO in higher concentrations.