Tlačiť
ESD occupational shoes, lace-up, Air Cushion, 32600/32610
Dodávateľ: Abeba
32600/35 32600/36 32600/37 32600/38 32600/39 32600/40 32600/41 32600/42 32600/43 32600/44 32600/45 32600/46 32600/47 32610/39 32610/40 32610/41 32610/42 32610/43 32610/44 32610/45 32610/46 32610/47 32610/38 32610/37 32610/36 32610/35
ABEB32600/35EA 99.08 EUR
ABEB32600/35 ABEB32600/36 ABEB32600/37 ABEB32600/38 ABEB32600/39 ABEB32600/40 ABEB32600/41 ABEB32600/42 ABEB32600/43 ABEB32600/44 ABEB32600/45 ABEB32600/46 ABEB32600/47 ABEB32610/39 ABEB32610/40 ABEB32610/41 ABEB32610/42 ABEB32610/43 ABEB32610/44 ABEB32610/45 ABEB32610/46 ABEB32610/47 ABEB32610/38 ABEB32610/37 ABEB32610/36 ABEB32610/35
ESD occupational shoes, lace-up, Air Cushion, 32600/32610
Obuv
Extremely light ESD shoe made from grained leather and fuel resistant PU sole.
  • Replaceable insole perspiration-absorbent lining
  • Sanitized® treatment reduces odour and bacterial growth
  • Air-cushioned PU sole with high slip resistance
Colour: White or black
Colour: White or black
Certifikácie: EN 61340 (ESD); EN ISO 20347:2012; O1, SRA.