Tlačiť
ESD occupational shoes, slip-on, Air Cushion, 32300/32310
Dodávateľ: Abeba
32300/35 32300/36 32300/37 32300/38 32300/39 32300/40 32300/41 32300/42 32300/43 32300/44 32300/45 32300/46 32300/47 32310/39 32310/40 32310/41 32310/42 32310/43 32310/44 32310/45 32310/46 32310/47
ABEB32300/35EA 96.27 EUR
ABEB32300/35 ABEB32300/36 ABEB32300/37 ABEB32300/38 ABEB32300/39 ABEB32300/40 ABEB32300/41 ABEB32300/42 ABEB32300/43 ABEB32300/44 ABEB32300/45 ABEB32300/46 ABEB32300/47 ABEB32310/39 ABEB32310/40 ABEB32310/41 ABEB32310/42 ABEB32310/43 ABEB32310/44 ABEB32310/45 ABEB32310/46 ABEB32310/47
ESD occupational shoes, slip-on, Air Cushion, 32300/32310
Obuv
Extremly light ESD shoe made from grained perforated white leather and a fuel resistant PU sole.
  • Replaceable insole
  • Sanitized® treatment reduces odour and bacterial growth
  • Air cushioned PU sole with high slip resistance
Colour: White or black
Certifikácie: EN 61340 (ESD); EN ISO 20347:2012; O1, SRA