Tlačiť
Eye showers for wall-mounting
Dodávateľ: KOEHLER TECHNISCHE PRODUKTEN
B-2011 B-2010
118-2191EA 1411.58 EUR
118-2191 118-2190
Eye showers for wall-mounting
Pomůcky první pomoci Sprchy
These eye showers with two shower heads are available with stainless steel or ABS plastic bowl.
  • Operated with hand plate
  • Quick action valve
  • Stainless steel pipes
Inlet: ½"
Outlet: 1¼"
Certifikácie: EN 15154 1+2