Tlačiť
Safety canisters
Dodávateľ: Köhler Technische Produkte
113-1247EA 126.69 EUR
113-1247 113-1246
Safety canisters
Nádoby bezpečnostní
Stainless steel safety canisters are ideal for transport of hazardous substances in public places.
  • Metallic flame barrier in filling/pouring opening
  • Grounding connection
Certifikácie: ADR/RID/IMDG approval (UN number).