Tlačiť
Occupational shoes, clogs, The Original, 7020/7021
Dodávateľ: Abeba
7020/39 7020/37 7020/41 7020/38 7020/43 7020/44 7020/46 7020/42 7020/45 7020/36 7020/40 7020/47 7020/35
ABEB7020/39EA 61.77 EUR
ABEB7020/39 ABEB7020/37 ABEB7020/41 ABEB7020/38 ABEB7020/43 ABEB7020/44 ABEB7020/46 ABEB7020/42 ABEB7020/45 ABEB7020/36 ABEB7020/40 ABEB7020/47 ABEB7020/35
Occupational shoes, clogs, The Original, 7020/7021
Obuv
Anti-static clogs made from smooth leather or perforated leather upper and a sole with cross/circle profile resistant to animal fats, oil and fuel.
  • Leather insole
  • Sanitized® treatment reduces odour and bacterial growth
  • Foldable and adjustable heel strap
Colour: White
Colour: White
Certifikácie: TÜV GS tested; EN ISO 20347:2012; OB, FO, SRC.