Tlačiť
ESD occupational shoes, clogs, Nature, 4040/4045/4050/4055
Dodávateľ: Abeba
4040/36 4040/37 4040/38 4040/39 4040/40 4040/41 4040/42 4040/43 4040/44 4040/45 4040/34 4040/35 4040/46 4040/47 4040/48 4045/34 4045/35 4045/36 4045/37 4045/38 4045/39 4045/40 4045/41 4045/42 4045/43 4045/44 4045/45 4045/46 4045/47 4045/48 4050/34 4050/35 4050/36 4050/37 4050/38 4050/39 4050/40 4050/41 4050/42 4050/43 4050/44 4050/45 4050/46 4050/47 4050/48 4055/43 4055/40 4055/36 4055/35 4055/34 4055/38 4055/39 4055/37 4055/48 4055/47 4055/46 4055/45 4055/44 4055/42 4055/41
114-2455EA 101.89 EUR
114-2455 114-2456 114-2457 114-2458 114-2459 114-2460 114-2461 114-2462 114-2463 114-2464 ABEB4040/34 ABEB4040/35 ABEB4040/46 ABEB4040/47 ABEB4040/48 ABEB4045/34 ABEB4045/35 ABEB4045/36 ABEB4045/37 ABEB4045/38 ABEB4045/39 ABEB4045/40 ABEB4045/41 ABEB4045/42 ABEB4045/43 ABEB4045/44 ABEB4045/45 ABEB4045/46 ABEB4045/47 ABEB4045/48 ABEB4050/34 ABEB4050/35 ABEB4050/36 ABEB4050/37 ABEB4050/38 ABEB4050/39 ABEB4050/40 ABEB4050/41 ABEB4050/42 ABEB4050/43 ABEB4050/44 ABEB4050/45 ABEB4050/46 ABEB4050/47 ABEB4050/48 ABEB4055/43 ABEB4055/40 ABEB4055/36 ABEB4055/35 ABEB4055/34 ABEB4055/38 ABEB4055/39 ABEB4055/37 ABEB4055/48 ABEB4055/47 ABEB4055/46 ABEB4055/45 ABEB4055/44 ABEB4055/42 ABEB4055/41
ESD occupational shoes, clogs, Nature, 4040/4045/4050/4055
Obuv
ESD shoe made from smooth or perforated leather with non foldable, adjustable heel strap and a rubber sole (resistant to fuels, oils, acids and bases).
  • Skin friendly leather coated, cork integrated insole with toe gripper
  • Sanitized® treatment reduces odour and bacterial growth
  • Slip resistant special rubber sole with shock-absorbent heel
  • Adjustable instep area with buckle fastening
Colour: White, black or dark blue
Colour: White, black or dark blue
Certifikácie: EN 61340 (ESD); TÜV GS DIN 4843/100 § 3.1v.8/93, EN ISO 20347:2012; OB, FO, A, SRC