Perfluorotetradecanoic acid 96%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
L13796.03
L13796.06
L13796.03EA
41.32
EUR
L13796.03
L13796.06
Perfluorotetradecanoic acid 96%
Perfluorotetradecanoic acid
Purity: 96%
CAS No.: 376-06-7
Molecular Formula: CF3(CF2)12CO2H
Formula Weight: 714.12
Melting Point: 130-132°
Boiling Point: 192°/60mm
UN No.: 3261
MDL No.: MFCD00153263
Vzorec:
C₁₄HF₂₇O₂ Teplota varu: 192 °C (60 mmHg) Teplota tavenia: 130…132 °C Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00153263 Číslo CAS: 376-06-7 EINECS: 206-803-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...