Tlačiť
Tubing, silicone, medical grade
Dodávateľ: DELTALAB
350034 350048 350612 350079 350912 350915 351622 352030 350810 351014 351420
228-4401EA 15.64 EUR
228-4401 228-4402 228-4403 228-4404 228-4406 228-4407 228-4408 228-4409 DELT350810 DELT351014 DELT351420
Tubing, silicone, medical grade
Hadičky Přenosová hadička
Medical grade silicone, translucent.
  • Excellent mechanical and chemical resistance
  • Autoclavable
For use in a variety of medical, pharmaceutical, industrial and clinical applications.
Temperature resistance: –50 to +200 °C
Hardness (Shore durometer A): 40 to 80