Tlačiť
ESD safety shoes, slip-on, X-Light, 7131032/7131037
Dodávateľ: Abeba
7131032/35 7131032/36 7131032/37 7131032/38 7131032/39 7131032/40 7131032/41 7131032/42 7131032/43 7131032/44 7131032/45 7131032/46 7131032/47 7131032/48 7131037/35 7131037/36 7131037/37 7131037/38 7131037/39 7131037/40 7131037/41 7131037/42 7131037/43 7131037/44 7131037/45 7131037/46 7131037/47 7131037/48
ABEB7131032/35EA 120.34 EUR
ABEB7131032/35 ABEB7131032/36 ABEB7131032/37 ABEB7131032/38 ABEB7131032/39 ABEB7131032/40 ABEB7131032/41 ABEB7131032/42 ABEB7131032/43 ABEB7131032/44 ABEB7131032/45 ABEB7131032/46 ABEB7131032/47 ABEB7131032/48 ABEB7131037/35 ABEB7131037/36 ABEB7131037/37 ABEB7131037/38 ABEB7131037/39 ABEB7131037/40 ABEB7131037/41 ABEB7131037/42 ABEB7131037/43 ABEB7131037/44 ABEB7131037/45 ABEB7131037/46 ABEB7131037/47 ABEB7131037/48
ESD safety shoes, slip-on, X-Light, 7131032/7131037
Obuv
ESD safety shoe made from smooth leather and fuel resistant PU sole.
  • Perspiration-absorbent special lining with silver ions
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Instep area with elastic band
Colour: White or black
Colour: White/black
Certifikácie: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC.