Tlačiť
Large Chairs and Stools, Vinyl
Dodávateľ: BIENS
4DL40GHLP01705 4DL40GBLU00505 4GM34GBLU00905 4GM34GBLU90737 4GM34GHLP02905 4GM34GHLP92737 4GM37GPLH00905 4GN10GBJU00905 4GN10GHJP02905 4GN17GPJH00905 4GN10GHJR02905 4DL40GDLP01505 4DL40GBLV00705
139-0210EA 578.82 EUR
139-0210 139-0211 BIEN4GM34GBLU00905 BIEN4GM34GBLU90737 139-1000 139-1001 BIEN4GM37GPLH00905 139-1002 139-0597 BIEN4GN17GPJH00905 BIEN4GN10GHJR02905 BIEN4DL40GDLP01505 BIEN4DL40GBLV00705
Large Chairs and Stools, Vinyl
Židle
Large ergonomic chairs, comfort and eco stools with high density foam and fire resistant vinyl coating.
  • 5-branch chromed, scuff resistant reinforced base
  • Soft and comfortable
  • The smooth watertight surface is easy to clean and decontaminate
  • Fire-resistant: M1 and EN 1021, 1-2
  • Gas lift column with shock absorber for height adjustment
Large chairs
Large seat and backrest with high density foam and resistant vinyl coating
Independently adjustable seat and backrest tilt with one fingertip lever thanks to an asynchronous mechanism (for asynchronous chairs)
Dimensions of the seat (W×D): 470×470 mm
Dimensions of the backrest (W×H): 400×420 mm
Stools
Round seat in M1 Ginkgo vinyl with high density soft foam
Diameter of the seat: 350 mm
Dimensions of backrest (W×H): 360×220 mm
Seat thickness: 600 mm for the Comfort model and 300 mm for the Eco one
Option: Backrest/Foot control height adjustment available on request
The Eco models are also available in black colour (on request)