Tlačiť
Biological safety cabinets, class II, BioVanguard Green Line
Dodávateľ: CLEANAIR BY BAKER
NH501003 NH501001 NH601001 NH401003 NH401001 NH301001 NH601003
135-1717EA 18065.78 EUR
135-1717 135-1715 135-1723 135-1709 135-1692 135-1705 135-1725
Biological safety cabinets, class II, BioVanguard Green Line
Digestoře Biologické boxy
BioVanguard Green Line is a Class II cabinet range which offers simplicity, robustness and high reliability and ensures the highest level of protection for the operator, product and environment, minimising the hazards inherent with working with agents assigned to biosafety levels one, two and three. It is designed for microbiological research with biological agents (e.g. bacteria, viruses, etc) and allergens.
  • Easy to install, easy to operate, easy to clean, easy to decontaminate and low maintenance
  • Microprocessor control with automatic compensation for filter clogging
  • User friendly LCD control panel, positioned in an angle, showing the operator all relevant functions and alarm (visual and audible) at a glance
  • Ergonomic epoxy coated aluminium arm rest contributes to comfortable working position of the operator
  • Two independent blowers and anemometers to control chamber airflow and inlet/exhaust flow
  • Exhaust and chamber HEPA filters H-14 of efficiency 99,995% MPPS (99,999% DOP)
  • RS232 and RS485 connection
The stainless steel worktop with V-shaped air slits provides superior safety by preventing blockages to the inflow and contamination of operator’s sleeves. The front window is laminated safety glass and can be lifted completely, opening up the entire work area for easy cleaning and/or (un)loading of large items.
BioVanguard is equipped as standard with:
- Epoxy coated exterior
- Stainless steel interior
- Stainless steel one-piece or segmented worktop
- Pre-filter and drip tray
- 2× electrical sockets
Available with manual or electrical windows.
BioVanguard Series is designed and manufactured according to international biosafety standards EN12469: Class II, NSF49:
Class II Type A2, JIS K 3800: Class II Type A2 and PIC’s. The working area is classified as ISO 14644: Class 5 and GMP
Annex 1: Class A.
Certifikácie: BioVanguard series is designed and manufactured according to international biosafety standards EN 12469: Class II, NSF49:
Class II Type A2, JIS K 3800: Class II Type A2 and PIC’s. The working area is classified as ISO 14644: Class 5 and GMP
Annex 1: Class A.