Tlačiť
PCR plates, 96-well, half skirt, Axygen®
Dodávateľ: Corning
PCR-96-HS-C PCR-96M2-HS-C PCR-96M2-HSR
731-1010EA 66.99 EUR
731-1010 732-0803 731-4124
PCR plates, 96-well, half skirt, Axygen®
Mikrodestičky Destičky pro PCR
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxin for highest sample integrity
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): ABI Fast PCR 5700, 7000, 7700, BioRad iCycler, MyiQ, iQ5, ABI sequencers.