Tlačiť
PCR plates, 384-well, Axygen®
Dodávateľ: Corning
PCR-384-C PCR-384M2-C PCR-384-RGD-C PCR-384M2CPBC
732-0906EA 79.82 EUR
732-0906 732-0666 732-1037 AXYGPCR-384M2CPBC
PCR plates, 384-well, Axygen®
Mikrodestičky Destičky pro PCR
PP plates, silicone mats. Designed to fit 384-well thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxin for highest sample integrity