Tlačiť
Cleanroom shirts, 7R011
Dodávateľ: FRISTADS KANSAS
1006355402XL 1006355403XL 100635540L 100635540M 100635540S 100635540XL 100635540XS
KWIN1006355402XLEA 77.42 EUR
KWIN1006355402XL KWIN1006355403XL KWIN100635540L KWIN100635540M KWIN100635540S KWIN100635540XL KWIN100635540XS
Cleanroom shirts, 7R011
Pracovní oděvy
These shirts with long sleeves are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres.
  • Bellows pleat in back
  • Concealed snap fasteners
  • Shirt collar with snap fastener
  • Two chest pockets with flap
  • Elasticated cuffs
The fibres are continuous filament fibres, which means that the garments do not emit particles or fibres. The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
Weight: 125 g/m²