Tlačiť
Safety ankle boots, lace-up, Steelite™ AQUA All Wheather Boots FW57
Dodávateľ: Portwest
FW57BKR47 FW57BKR46 FW57BKR45 FW57BKR44 FW57BKR43 FW57BKR42 FW57BKR41 FW57BKR40 FW57BKR39
PRTWFW57BKR47EA 137.99 EUR
PRTWFW57BKR47 PRTWFW57BKR46 PRTWFW57BKR45 PRTWFW57BKR44 PRTWFW57BKR43 PRTWFW57BKR42 PRTWFW57BKR41 PRTWFW57BKR40 PRTWFW57BKR39
Safety ankle boots, lace-up, Steelite™ AQUA All Wheather Boots FW57
Obuv
Anti-static, waterproof safety ankle boot manufactured from nubuck leather upper plus a dual-density PU/TPU sole, resistant to oil and heat (200 °C).
  • Waterproof, breathable lining
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
For outdoor use.
Colour: Black.
Certifikácie: EN ISO 20345:2011; S3, SRC, HRO.