Tlačiť
Safety shoes, lace-up, Steelite™ WORK Thor FW44
Dodávateľ: Portwest
FW44BKR47 FW44BKR46 FW44BKR44 FW44BKR43 FW44BKR42 FW44BKR41 FW44BKR40 FW44BKR39 FW44BKR38
PRTWFW44BKR47EA 79.02 EUR
PRTWFW44BKR47 PRTWFW44BKR46 PRTWFW44BKR44 PRTWFW44BKR43 PRTWFW44BKR42 PRTWFW44BKR41 PRTWFW44BKR40 PRTWFW44BKR39 PRTWFW44BKR38
Safety shoes, lace-up, Steelite™ WORK Thor FW44
Obuv
Anti-static safety shoes made from water resistant full grain leather and a dual-density PU/TPU sole, resistant to oil.
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifikácie: EN ISO 20345:2011; S3, SRC.