Tlačiť
Glove liners, Camapur® Comfort Antistatik 624+
Dodávateľ: KCL
624.10 624.6 624.7 624.8 624.9
KCHL624.10EA 0 EUR
KCHL624.10 KCHL624.6 KCHL624.7 KCHL624.8 KCHL624.9
Glove liners, Camapur® Comfort Antistatik 624+
These woven polyamide gloves with copper thread and PU coated fingertips fulfill all requirements for the classification as anti static (ESD) gloves.
  • Very breathable, good sensitivity, high dexterity
  • Roughened PU-coating for good grip
  • Good antistatic and electrostatic properties
  • Washable
  • Different colours of knit wrist band indicate the size
Ideal for usage in assembly works, electrical and automotive industries, chip processing and precision electronics.
Thickness: 0,6 ± 0,1 mm
Colour: Grey (Wristband in different colours)
EN 1149-1/2, EN 388 (2130); Cat. II
Certifikácie: EN 388:2016 (113XX), EN 16350:2014-07; Cat. II